tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:286961-14-6
:
Formula:C₁₆H₂₈BNO₄
MDL Number:MFCD03840345
:309.21 g/mol
Melting Pt:116 °C
:GHS07
Storage Temperature:Omgivande