Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxy-3-pyridineboronic acid pinacol ester
2-Methoxy-3-pyridineboronic acid pinacol ester 97%
2-Methoxy-3-pyridineboronic acid pinacol ester 97%
2-Methoxy-3-pyridineboronic acid pinacol ester 97%
APOSOR360013-5G
:
Apollo Scientific
:
2-Methoxy-3-pyridineboronic acid pinacol ester 97%
APOSOR360013-5G
:
Apollo Scientific
:
Förpackningstyp:
Bottle
Förp.:
5 g
Seq:
3173
:
:
235.09 g/mol
:
Omgivande
:
532391-31-4
:
C₁₂H₁₈BNO₃
:
MFCD05663507