Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Nicotinanilide
Nicotinanilide 95%
Nicotinanilide 95%
Nicotinanilide 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Nicotinanilide 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1752-96-1
:
Formula:
C₁₂H₁₀N₂O
MDL Number:
MFCD00023572
:
198.23 g/mol
Melting Pt:
119…120 °C
:
GHS07
Storage Temperature:
Omgivande