3-Ethoxy-4-methoxybenzaldehyde
Catalog # APOSOR25717-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:180.20 g/mol
Boiling Pt:155 °C/9,8mmHg
Melting Pt:51…53 °C
Flash Pt:>113 °C
:GHS07
Storage Temperature:Omgivande
CAS Number:1131-52-8
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00010128