3-Aminoisonicotinamide
Catalog # APOSOR2138-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:5 g
- Seq:3173
Specifications
About this item
:
:137.14 g/mol
Melting Pt:84…86 °C
:GHS07
Storage Temperature:Kylskåp
CAS Number:64188-97-2
Formula:C₆H₇N₃O
MDL Number:MFCD08235178