Ethyl tricosanoate
Catalog # APOSOR111030-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:382.67 g/mol
Storage Temperature:Omgivande
CAS Number:18281-07-7
Formula:C₂₅H₅₀O₂