
Ytterbium(III) chloride, vattenfri 99,9%
Catalog # APOSIN3796-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:10 g
- Seq:3236
Specifications
About this item
:
:279.4 g/mol
Boiling Pt:1900 °C
Melting Pt:703 °C
Density:4,06 g/cm³ (25 °C)
:GHS07
Storage Temperature:Omgivande
CAS Number:10361-91-8
Formula:Cl₃Yb
MDL Number:MFCD00011288