ContactUsLinkComponent
Tin(IV) sulphide 99,9%
Irritant
Tin(IV) sulphide 99,9%
Catalog # APOSIN3676-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Tin(IV) sulphide 99,9%
Catalog # APOSIN3676-5G
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Förpackningstyp:
    Bottle
  • Förp.:
    5 g
  • Seq:
    3173
Specifications
About this item
 :
 :182.84
Boiling Pt:600 °C
Density:4,5 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Omgivande
CAS Number:1315-01-1
Formula:S₂Sn
MDL Number:MFCD00049546