ContactUsLinkComponent
Tenntetraklorid 99.99%
Corrosive
Tenntetraklorid 99.99%
Catalog # APOSIN3649-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Tenntetraklorid 99.99%
Catalog # APOSIN3649-25G
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Förpackningstyp:
    Bottle
  • Förp.:
    25 g
  • Seq:
    3308
Specifications
About this item
 :
Boiling Pt:114 °C
Melting Pt:–33 °C
Density:2,226 g/cm³ (25 °C)
Storage Temperature:Omgivande
ADR:8,II
 :260.52 g/mol
EINECS:231-588-9
UN:1827
 :GHS05
CAS Number:7646-78-8
Formula:Cl₄Sn
MDL Number:MFCD00011242