
Nickel(II) dibutyldithiocarbamate
Catalog # APOSIN2708-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:25 g
- Seq:3308
Specifications
About this item
:
:467.43 g/mol
Melting Pt:89 °C
:GHS07
Storage Temperature:Omgivande
CAS Number:13927-77-0
Formula:C₉H₁₉NS₂
MDL Number:MFCD00067273