Gallium tribromide, vattenfri 99,9%
Catalog # APOSIN1864-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:279 °C
:309.44 g/mol
Storage Temperature:Omgivande
CAS Number:13450-88-9
Formula:Br₃Ga
MDL Number:MFCD00016100