
Cerium i mineralolja 99,9%, chips
Catalog # APOSIN1399-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förp.:25 g
- Seq:3308
Specifications
About this item
:
:140.12 g/mol
Boiling Pt:3468 °C
Melting Pt:795 °C
Density:6,67 g/cm³ (25 °C)
Storage Temperature:Omgivande
EINECS:231-154-9
UN:3078
ADR:4.3,II
:GHS07
CAS Number:7440-45-1
Formula:Ce
MDL Number:MFCD00010924