Order Entry
Sweden
Orders LinkContactUsLinkComponent
Bismuth karbonat basic 99.99%

Bismuth karbonat basic 99.99%

Supplier:   Apollo Scientific
Bismuth karbonat basic 99.99%
Supplier:  Apollo Scientific
Certificates
Bismuth karbonat basic 99.99%

2 Options Available Below

Product Details & Documents
Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Density:6,86 g/cm³ (25 °C)
  • Storage Temperature:Omgivande
  • EINECS:227-567-9
Product Family Options

Product Information

  • Förpackningstyp
    Förp.
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 4 100,00 kr
    4 100,00 kr
    1 * 100 g
       4 100,00 kr

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 700,00 kr
    1 700,00 kr
    1 * 25 g
       1 700,00 kr