Propynol ethoxylate
Catalog # APOSBIPG1713-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:100.12 g/mol
Storage Temperature:Omgivande
CAS Number:3973-18-0
Formula:C₅H₈O₂
MDL Number:MFCD00099294