Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitrophenyl palmitate
4-Nitrophenyl palmitate
4-Nitrophenyl palmitate
APOSBIN0442-10G
:
Apollo Scientific
:
4-Nitrophenyl palmitate
APOSBIN0442-10G
:
Apollo Scientific
:
Förpackningstyp:
Bottle
Förp.:
10 g
Seq:
3236
:
:
377.52 g/mol
:
GHS07
:
Frys
:
1492-30-4
:
C₂₂H₃₅NO₄
:
MFCD00047732