
Roquefortine C
Catalog # APOSBIBR1088-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 mg
- Seq:2070
Specifications
About this item
:
:389.5 g/mol
Storage Temperature:Frys
UN:2811
ADR:6.1,II
:GHS06
CAS Number:58735-64-1
Formula:C₂₂H₂₃N₅O₂