Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
α-Amanitin
α-Amanitin (from Amanita phalloides)
α-Amanitin (from Amanita phalloides)
Toxic
Health
α-Amanitin (from Amanita phalloides)
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
Health
α-Amanitin (from Amanita phalloides)
Supplier:
Apollo Scientific
About this item
CAS Number:
23109-05-9
:
Melting Pt:
254…255 °C
Storage Temperature:
Frys
EINECS:
245-432-2
UN:
3462
ADR:
6.1,II
:
GHS06
:
GHS08