Chemical resistant tape, Kappler™ ChemTape®
NEWPWPL591
: NEW PIG
- Bredd:50 mm
- Beskrivning:Chemical resistant tape
- Längd:55 m
- Förp.:1 Roll
Seam-sealing chem tape designed and tested for use with chemical-resistant clothing.
- Tears easily by hand
- Protects better than duct tape
- Excellent for securing gloves and boots to protective clothing
Ideal for haz-mat response, this tape is designed and tested to ensure chemical resistance.