PIG® HazMat Spill kit in see-thru duffel bag
Catalog # NEWPKITE389
Supplier: NEW PIG
Specifications
- Beskrivning:Spill kit, includes 40 mat pads, 3 socks, 4 pillows and 5 disposal bags
- B×D×H:280×510×230 mm
- Absorptionsförmåga:27,4 L
- Förp.:1
Specifications
About this item
This portable PIG® spill kit is compact to fit in tight spaces, and the clear top makes it easy to verify contents during inspections or spill response.
- Use socks to confine a spill before it can spread; use mats and pillows to soak it up
- Absorbents are specially treated to absorb high concentrations of corrosive liquids, such as 98% sulfuric acid and 30% sodium hydroxide, better than any other absorbent in the market
- Clear plastic top flap lets you easily check contents, so you know you’re prepared prior to a spill or an audit
- Heavy-duty, black vinyl duffel bag stands up to wear and tear; extra pockets let you store additional tools and supplies
- Bold Spill Kit lettering for quick identification
- Includes temporary disposal bags for used absorbents
The compact size is ideal for storage in fleet vehicles, at individual workstations or in other tight spaces.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.