Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Trifluoromethylcatechol
3-Trifluoromethylcatechol 95%
3-Trifluoromethylcatechol 95%
3-Trifluoromethylcatechol 95%
APOSPC910273-1G
:
Apollo Scientific
:
3-Trifluoromethylcatechol 95%
APOSPC910273-1G
:
Apollo Scientific
:
Förpackningstyp:
Bottle
Förp.:
1 g
Seq:
3047
:
:
178.109 g/mol
:
Omgivande
:
113678-92-5
:
C₇H₅F₃O₂
:
MFCD18395727