Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
4-(Difluoromethoxy)-2-fluoroaniline
4-(Difluoromethoxy)-2-fluoroaniline
4-(Difluoromethoxy)-2-fluoroaniline
4-(Difluoromethoxy)-2-fluoroaniline
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Difluoromethoxy)-2-fluoroaniline
Supplier:
Apollo Scientific
About this item
CAS Number:
1003865-65-3
:
Formula:
C₇H₆F₃NO
MDL Number:
MFCD14698443
:
177.124 g/mol
:
GHS07
Storage Temperature:
Kylskåp