0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
2-Fluoroisoniazide
2-Fluoroisoniazide
2-Fluoroisoniazide
:
Apollo Scientific
2-Fluoroisoniazide
:
Apollo Scientific
:
369-24-4
:
:
C₆H₆FN₃O
:
MFCD08689558
:
155.13 g/mol
:
GHS07
:
Omgivande