4-Methylheptafluorotoluene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:778-35-8
:
Formula:C₈H₃F₇
MDL Number:MFCD00012173
:232.098 g/mol
Boiling Pt:143…145 °C
Density:1,528 g/cm³ (25 °C)
Storage Temperature:Omgivande
UN:1993
ADR:3,III
:GHS02
:GHS07