Order Entry
Sweden
ContactUsLinkComponent
4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one
4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one
Catalog # APOSPC400599-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one
Catalog # APOSPC400599-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Förpackningstyp:
    Bottle
  • Förp.:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :247.247 g/mol
Storage Temperature:Omgivande
CAS Number:143491-54-7
Formula:C₈H₁₀FN₃O₃S
MDL Number:MFCD00870151