Order Entry
Sweden
ContactUsLinkComponent
 
Irritant
6-[(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-trihydroxypiperidin-1-yl]hexanoic acid
Catalog # APOSOR8120T-5MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-[(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-trihydroxypiperidin-1-yl]hexanoic acid
Catalog # APOSOR8120T-5MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Förpackningstyp:
    Bottle
  • Förp.:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :277.314 g/mol
Melting Pt:185…187 °C
 :GHS07
Storage Temperature:Frys
CAS Number:79206-51-2
Formula:C₁₂H₂₃NO₆
MDL Number:MFCD01320383