ContactUsLinkComponent
5-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-2-furaldehdye
Irritant
5-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-2-furaldehdye
Catalog # APOSOR8086-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-2-furaldehdye
Catalog # APOSOR8086-250MG
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Förpackningstyp:
    Bottle
  • Förp.:
    250 mg
  • Seq:
    2590
Specifications
About this item
 :
 :255.226 g/mol
Melting Pt:125…128 °C
 :GHS07
Storage Temperature:Omgivande
CAS Number:116750-06-2
Formula:C₁₄H₉NO₄
MDL Number:MFCD02180376