Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
5-Isopropylcyclohexane-1,3-dione hydrate
5-Isopropylcyclohexane-1,3-dione hydrate 97%
5-Isopropylcyclohexane-1,3-dione hydrate 97%
Irritant
5-Isopropylcyclohexane-1,3-dione hydrate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Isopropylcyclohexane-1,3-dione hydrate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
207399-18-6
:
Formula:
C₉H₁₄O₂·H₂O
MDL Number:
MFCD00150873
:
172.222 g/mol
Melting Pt:
85…87 °C
:
GHS07
Storage Temperature:
Omgivande