(3S)-3-{[(Pyridin-2-yl)methyl]amino}piperidine, N1-BOC protected
Catalog # APOSOR62011-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:291.389 g/mol
Storage Temperature:Omgivande
CAS Number:1349702-31-3
Formula:C₁₆H₂₅N₃O₂
MDL Number:MFCD19381686