Order Entry
Sweden
ContactUsLinkComponent
Tin(IV) chloride hydrate 98%
Corrosive
Tin(IV) chloride hydrate 98%
Catalog # APOSIN3650-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Tin(IV) chloride hydrate 98%
Catalog # APOSIN3650-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Förpackningstyp:
    Bottle
  • Förp.:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :260.50 g/mol (anhy)
Storage Temperature:Omgivande
UN:2440
ADR:8,III
 :GHS05
CAS Number:7646-78-8
 :10026-06-9
Formula:SnCl₄·xH₂O
MDL Number:MFCD00216885