Thiol-PEG3-Alcohol
Catalog # APOSBIPG1840-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
:166.239 g/mol
Storage Temperature:Omgivande
CAS Number:56282-36-1
Formula:C₆H₁₄O₃S
MDL Number:MFCD04116140