Cholesteryl phosphorylethanolamine
Catalog # APOSBIB6033-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:100 mg
- Seq:2440
Specifications
About this item
:
:509.701 g/mol
Storage Temperature:Frys
CAS Number:120090-10-0
Formula:C₂₉H₅₂NO₄P
MDL Number:MFCD09841711