TG1 electroporation-competent cells
Catalog # AGLS200123
Leverantör: AGILENT
Specifikationer
- Beskrivning:TG1 electroporation-competent cells
- Pk Info:5×0,1 ml (each)
- Funktion:For use in phage display library preparation, ≥1×10¹⁰ transformations/µg
- Förp.:5
Specifikationer
Om denna produkt
Phage display libraries from electroporation-competent cells with high transformation efficiency.
- Just thaw, combine with DNA, and electroporate to get started quickly
- Efficiency of ≥1×10¹⁰ transformations/µg of pUC18 DNA
- Kit size 5×0,1 ml
Electroporation-competent cells capable of producing greater than 1×10¹⁰ transformants/µg of DNA. These electroporation-ready cells need only be thawed, mixed with DNA, and electroporated. TG1 electroporation-competent cells are recommended for preparation of phage display libraries.