EletroTen-blue electroporation-competent cells
Catalog # AGLS200159
Supplier: AGILENT
Specifications
- Beskrivning:EletroTen-Blue electroporation-competent cells
- Pk Info:5×0,1 ml
- Funktion:Highest electroporation efficiency (Hee) phenotype, use for extrememly demanding cloning, 3≥3×10¹⁰ transformants/µg
- Förp.:5
Specifications
About this item
Achieve the highest transformation efficiencies available when cloning large plasmids and ligated DNA.
- Separate DNA fragments and libraries up to 5000 bp in 1 to 2 min/sample
- Achieve objective quantitative and purity measurements from as little as 2 µl of DNA, sensitive down to 5 pg/µl
- Ideal for sample QC in NGS workflows
- Partially used ScreenTape devices can be saved and used at a later time
- No need to batch samples
Agilent EletroTen-Blue Electroporation-Competent Cells offer the highest transformation efficiencies available, dramatically improving cloning large plasmids and ligated DNA. These cells also offer the highest electroporation efficiency (Hee) phenotype, which confers increased resilience in harsh electroporation conditions.
Caution: For research use only. Not for use in diagnostic procedures.