Sulphochlorophenol S sodium calcium salt, Sigma-Aldrich®
Catalog # S5752-500MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Förp.:500 mg
- Seq:2725
Specifications
About this item
:
:789.53 g/mol
Storage Temperature:Kylskåp
CAS Number:108321-09-1
Formula:C₂₂H₁₄Cl₂N₄O₁₆S₄
MDL Number:MFCD00151404