Order Entry
Sweden
ContactUsLinkComponent
FURA-2 pentapotassium salt, Sigma-Aldrich®
FURA-2 pentapotassium salt, Sigma-Aldrich®
Catalog # F0763-1MG
CAS Number:  
undefined
FURA-2 pentapotassium salt, Sigma-Aldrich®
Catalog # F0763-1MG
CAS Number:  

Specifications

  • Förp.:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :832 g/mol
Storage Temperature:Frys
CAS Number:113694-64-7
Formula:C₂₉H₂₂K₅N₃O₁₄