Antimicrobial foam hand cleanser, OxyBAC®
Catalog # EVONOXY1LBG
Leverantör: Deb-STOKO
Specifikationer
- Beskrivning:Foam hand cleanser with 1 L cartridge
- Förp.:6
Specifikationer
Om denna produkt
Antimicrobial rich-cream foam hand wash, perfume-free and dye-free.
- Contains accelerated hydrogen peroxide
- Skin hypoallergenic
- Suitable for food facilities and healthcare workers
- Excellent skin cleansing
- Hygienic cartridge
Dispenses foam on the hands which is instantly ready for washing and quick & easy rinsing from hands and sinks. Helps to prevent drying and leave the skin feeling smooth after use.
Certifications: HACCP Certified. Passes EN1499.
Caution: Use biocides safely. Always read the label and product information before use.