Disinfectant sprays, Des E, SoftCare™
Catalog # DIVY101104048
Supplier: Diversey
Specifications
- Beskrivning:Soft Care Des E spray liquid hand disinfectant, 20 L
- Seq:03
- Förp.:1 Bottle
Specifications
About this item
These SoftCare™ Des E disinfectant sprays, alcohol-based are effective against bacteria, virus and fungi. It is made up of 70% ethanol, which ensures instant hand hygiene.
- Leave on product, so does not require water
- Prevents infection
- Frangrance free
- Dye free
This disinfectant spray is ideal for use in healthcare and food processing areas.Liquid format suitable for spray application.
Caution: Use biocides safely. Always read the label and product information before use.