Sterilisation wraps, sequential, KC100 KIMGUARD™
Katalog # KIMB10712
Leverantör: KIMBERLY CLARK
Specifikationer
- Färg:Ljusblå
- Beskrivning:Sterilisation wrap, Kimguard® KC300
- Mått:305×305 mm
- Seq:01
- Environmentally Preferable:Waste Reducing
- Förp.:1.000
Specifikationer
Om denna produkt
Available in six strengths, from KC100 to KC600, to offer precisely the right protection for your needs
KIMGUARD™ KC100 Sterilization Wraps are the first of six strength levels. Printed with its name, size, and lot code.
Choosing is easy; the higher the number, the stronger the wrap. Sequential wrap. Used to package linen towels, packs, and gowns, light basin sets, and single instruments for sterilization. Low-lint fabric delivers barrier and strength properties that help prevent contamination after sterilization. Steam and EO compatible. Naturally latex-free. Non sterile. Multiple sizes.
Ordering information: Case count indicates the number of sheets included.