Cut resistant gloves, JACKSON SAFETY* G60, Level 3
A range of cost effective, durable gloves and sleeves suitable for processes where there is cutting or laceration risk to workers hands and arms.
- Latex free
- Available in different sizes with colour coded cuff
- Polyurethane coating offering good dexterity, dry and oil grip
These gloves are suitable for metal fabrication, glass and sharp objects handling, machinery and bending
small parts.
Level 4 abrasion resistance with high breathability as material transmits heat away. Durable and comfortable.
: Level 3 cut resistance (EN 388:4342). EN 420 standard protection to wrist area.