ANTI-NTRK3 Mouse Monoclonal Antibody [clone: BS337]
Katalog # BSENM-1837-100
Leverantör: Biosensis
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:NT-3 growth factor receptor
- Antigensymbol:NTRK3
- Klonalitet:Monoclonal
- Klon:BS337
- Konjugation:Unconjugated
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG2b
- Reaktivitet:Human,Rat,Mouse
- Epitop:Extracellular, amino acids 397-408 of human TrkC
- Korsad adsorption:No
- Form:Lyophilized
- Gen-ID:Q16288 (NTRK3_HUMAN)
- Antigen synonymer:Trk-C|TrkC tyrosine kinase|Neurotrophic tyrosine kinase receptor type 3|GP145-TrkC
- Aminosyretal:397 to 408
- Förvaringstemperatur:Store lyophilized antibody at 2-8C. After reconstitution divide into aliquots and store at -20C for long-term storage. Store at 2-8C short-term (up to 4 weeks). Avoid repetitive freeze/thaw cycles.
- Shipping temperature:RT
- Immunogen:Synthetic peptide immunogen, KEPFPESTDNFI
- Rening:Protein G purified
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Receptor tyrosine kinase involved in nervous system and probably heart development. Upon binding of its ligand NTF3/neurotrophin-3, NTRK3 autophosphorylates and activates different signaling pathways, including the phosphatidylinositol 3-kinase/AKT and the MAPK pathways, that control cell survival and differentiation (Ref: uniprot.org).
Applications: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde.
Immunocytochemistry (1-2 ug/mL): Tested on fixed (4% formaldehyde) human cells.
Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.