Medium tape measure sleeve, DBI-SALA, 3M™
Catalog # MMMA1500099
Leverantör: 3M
Specifikationer
- Beskrivning:Tape measure sleeve
- Förp.:1
Specifikationer
Om denna produkt
Tape measure holster.
- Adjusts to virtually any wrist size with hook and loop system
- Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
- Elastic band helps ensure a comfortable fit
- Attached trigger can be used to quickly work with many hand tools
The DBI-SALA® Tape measure sleeve conforms to the size of virtually any tape measure and allows the tool to be safely tethered while working at height. The sleeve is constructed from a heavy-duty neoprene material and is designed to be used with the Tape measure retractor holster (sold separately), or on its own using a belt clip on the back of the sleeve. A belt clip on the back of the sleeve allows the sleeve to be holstered quickly. A D-ring on the sleeve allows the user to tie off their tape measure.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.