VWR® Traceable® Replacement Platinum RTD Stainless-Steel Probe
620-2810
- Mätområde:‒100...200 °C
- Beskrivning:VWR® Traceable® replacement platinum RTD stainless-steel probe for RTD thermometer (VWRI620-1981)
- Längd:10' PTFE cable
- Förp.:1
- Typ:Platinum RTD
Optimise performance of thermometer.
- Replace the probe or keep a convenient spare
Replacement probe for RTD thermometer (VWRI620-1981).