Cutting tool, WebSeal™, chromacol
Katalog # 548-0233
Leverantör: CHROMACOL
Specifikationer
- Beskrivning:WebSeal™ skärverktyg
- Seq:40
- Förp.:1
Specifikationer
Om denna produkt
This cutting tool allows removal of individual vials as a sealed unit, if required.