Sterilin® Lids for 96-well Microtitre Plates
Katalog # 634-0011
Leverantör: STERILIN
Specifikationer
- Sterilitet:Sterilt
- Färg:Klar
- Beskrivning:Lid for 96-well plate
- Förp.:50
Specifikationer
Om denna produkt
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
- Manufactured from virgin polystyrene for excellent optical clarity
- Manufactured in ISO Class 7 Clean Room without surface treatment for a medium binding surface
- Available irradiated for sterility and a more hydrophilic surface, providing high binding
- Suitable for use at a wavelength of 340 nm
- Minimizes the risk of contamination from surrounding wells with condensation rings on the lid raised well rims on the base
- Ensures even temperature distribution around each well with a hanging well design
Delivery: Supplied without lid; may be purchased separately.