General purpose gloves, A020
Catalog # PRTWA020WHRL
Supplier: Portwest
Specifications
- Handskstorlek:L
- Ej handspecifika/handspecifika:Handspecifika
- Engångs/Återanvändbar:Återanvändbar
- Färg:Vit
- Seq:03
- Modell:A020
- Kragmodell:Stickad mudd
- Mätare:13
- Förp.:960
Specifications
About this item
These gloves are excellent for jobs requiring high dexterity and perfect for intricate tasks.
- Maximum tactile feedback
- Seamless 13 gauge liner
- Low linting construction for minimal contamination
Made of pylon.
Certifications: CE Cat 1