Maximum cut protection nitrile foam gloves, A645
Supplier: Portwest
Certificates
About this item
Nitrile foam coating in the gloves provides excellent grip in wet and dry conditions. Bright green liner provides easy identification.
- Level D cut resistance
- Green - High risk cut protection
- Palm dipped to increase dexterity and ventilation
- Protects against cuts and abrasion
- Retail bag which aids presentation for retail sales
- 13 gauge liner for a perfect fit
- 100% breathable seamless liner
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level A4. EN 388:2003 - 4.5.4.4. EN 388:2016 - 4.X.4.4.D. EN 420. CE Food Safe. EN cut protection levels.
Specifications
- Catalog No:
- PRTWA645E8RS
- PRTWA645E8RL
- PRTWA645E8RXL
- PRTWA645E8RM
- Modell:
- A645
- A645
- A645
- A645
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Material:
- Nitril
- Nitril
- Nitril
- Nitril
- Yta:
- Palm dipped
- Palm dipped
- Palm dipped
- Palm dipped
- Mätare:
- 13
- 13
- 13
- 13
- Godkända för livsmedelskontakt:
- +
- +
- +
- +