Classic Essentials Rain Suits, L440
Leverantör: Portwest
Certifikat
Om denna produkt
Waterproof jacket and pants with taped seams. Vented back yoke and eyelets for added breathability and comfort with pack away hood for added functionality.
- Front zip opening for easy access
- 2 pockets for secure storage
- Elasticated cuffs for a secure fit
- Stud adjustable hems for a secure fit
- Fully elasticated waistband for ultimate wearer comfort
Shell fabric : 100% polyester, 190T, PVC coated 190 g.
Certifications: Standards: EN 343:2019 Class 3:1 X (WP 5,000mm)
Specifikationer
- Catalog No:
- PRTWL440NARXL
- PRTWL440NARXXXL
- PRTWL440NARM
- PRTWL440NARL
- PRTWL440NARS
- PRTWL440NARXXL
- Modell:
- L440
- L440
- L440
- L440
- L440
- L440
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester, 190T, PVC coated
- 100% polyester, 190T, PVC coated
- 100% polyester, 190T, PVC coated
- 100% polyester, 190T, PVC coated
- 100% polyester, 190T, PVC coated
- 100% polyester, 190T, PVC coated
- Vikt:
- 190 g
- 190 g
- 190 g
- 190 g
- 190 g
- 190 g