Fleece jackets, Aran ladies F282
Supplier: Portwest
Certificates
About this item
The Aran Ladies Fleece provides a stylish and flattering fit. It is the perfect addition to any workwear wardrobe.
- Made from a soft lightweight 100% polyester
- Anti pill finish, high comfort and durability
- All-weather protection
- Four zipped pockets for ample storage
- Drawcord adjustable hem
- Weight: 280 g
Varying fabric weights offer versatility and choice across the range and ensure there is a garment to suit every need from extra layering to total outdoor protection.
Certifications: EN 14058 Class 1.
Specifications
- Catalog No:
- PRTWF282WHRL
- PRTWF282WHRM
- PRTWF282WHRS
- PRTWF282WHRXL
- PRTWF282WHRXS
- PRTWF282WHRXXL
- Modell:
- F282
- F282
- F282
- F282
- F282
- F282
- Outfit applikationer:
- Casual, Cleanroom, General Purpose
- Casual, Cleanroom, General Purpose
- Casual, Cleanroom, General Purpose
- Casual, Cleanroom, General Purpose
- Casual, Cleanroom, General Purpose
- Casual, Cleanroom, General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Kön:
- För damer
- För damer
- För damer
- För damer
- För damer
- För damer
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- Vikt:
- 580 g
- 560 g
- 540 g
- 620 g
- 520 g
- 640 g