High visibility waistcoats, MeshAir band and brace, C370
Leverantör: Portwest
Certifikat
Om denna produkt
This full mesh vest, uses our MeshAir fabric which allows increased airflow to keep the wearer cool. A perfect choice for those working in warmer environments.
- Cooling mesh fabric for increased breathability
- Reflective tape for increased visibility
- Hook and loop closure for easy access
- Generous fit for wearer comfort
Shell fabric: MeshAir Hi-Vis, 150 g.
Certifications: RIS-3279-TOM ISSUE 1 (orange only); EN ISO 20471 Class 2
Caution: Do not dry clean.
Specifikationer
- Catalog No:
- PRTWC370YERL/XL
- PRTWC370YERS/M
- PRTWC370ORRXX/3X
- PRTWC370ORR4X/5X
- PRTWC370ORRS/M
- PRTWC370ORRL/XL
- PRTWC370YER4X/5X
- PRTWC370YERXX/3X
- Modell:
- C370
- C370
- C370
- C370
- C370
- C370
- C370
- C370
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Stängningsstorlek:
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Tyg:
- MeshAir
- MeshAir
- MeshAir
- MeshAir
- MeshAir
- MeshAir
- MeshAir
- MeshAir
- Vikt:
- 205 g
- 183 g
- 245 g
- 245 g
- 183 g
- 205 g
- 245 g
- 245 g