Face shield, WP98 series
Catalog # MMMAWP98
Supplier: 3M
Specifications
- Beskrivning:WP98 series face shield
- Förp.:1
Specifications
About this item
The WP98 Series Face Shield is a high impact resistant, clear acetate faceshield designed to fit 3M hard hats. It can be used with the H8 Headgear easy faceshield replacement system.
- Provides protection against impacts, liquids, and chemical splashes
- Clear acetate faceshield
- High impact resistance
- Can be used with the H8 Headgear easy faceshield replacement system
- The shield goes inside the attachment for extra support
The WP98 Series clear acetate faceshield protects against high speed particles with low energy impact (0,86 g ball at 45 m/s), marking F. WP98 is certified for use with SecureFit helmets using holder U5B and is certified for use with G3000 and G3501 using holder U3-E. It is also certified for use with H8 Headgear.